Learn More
Cayman Chemical Myricetin 3-galactoside, Purity: ≥98%, CAS: 15648-86-9, 10 mg
SDP
Catalog No. 502911505
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits xanthine oxidase in a cell-free assay at 100 µg/ml; cytotoxic to K562 cells (IC50 = 220 µg/ml); inhibits hydrogen peroxide-induced MDA formation in K562 cells at 110, 220, and 440 µg/ml; reduces carrageenan-induced paw edema in wild-type mice at 0.26 and 0.78 mg/kg; reduces formalin-induced paw licking in mice
Catalog No. 50-291-1505 Supplier Cayman Chemical Supplier No. 3646710 MG
A flavonoid with diverse biological activities; inhibits xanthine oxidase in a cell-free assay at 100 µg/ml; cytotoxic to K562 cells (IC50 = 220 µg/ml); inhibits hydrogen peroxide-induced MDA formation in K562 cells at 110, 220, and 440 µg/ml; reduces carrageenan-induced paw edema in wild-type mice at 0.26 and 0.78 mg/kg; reduces formalin-induced paw licking in mice