Learn More
Medchemexpress LLC Myricetin 3'-glucoside | 520-14-9 | 1 MG
SDP
Catalog No. 5000417340
Encompass_Preferred
Myricetin 3'-glucoside is a glycoside derivative of quercetin. It significantly prevents ethanol-induced hepatotoxicity by reducing hepatic transaminase activity and inflammatory response in HepG2 cells, demonstrating a protective effect on alcohol-induced liver injury.
  • Glycoside derivative of quercetin
  • Prevents ethanol-induced hepatotoxicity
  • Reduces hepatic transaminase activity
  • Reduces inflammatory response in HepG2 cells
  • Protective effect on alcohol-induced liver injury
Catalog No. 50-004-17340 Supplier Medchemexpress LLC Supplier No. HYN79051MG
Myricetin 3'-glucoside is a glycoside derivative of quercetin. It significantly prevents ethanol-induced hepatotoxicity by reducing hepatic transaminase activity and inflammatory response in HepG2 cells, demonstrating a protective effect on alcohol-induced liver injury.
  • Glycoside derivative of quercetin
  • Prevents ethanol-induced hepatotoxicity
  • Reduces hepatic transaminase activity
  • Reduces inflammatory response in HepG2 cells
  • Protective effect on alcohol-induced liver injury