Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricetin 3'-glucoside | 520-14-9 | 5 MG
SDP

Catalog No. 5000418743
Encompass_Preferred

Myricetin 3'-glucoside is a glycoside derivative of quercetin. It significantly prevents ethanol-induced hepatotoxicity by reducing hepatic transaminase activity and inflammatory response in HepG2 cells, indicating a protective effect on alcohol-induced liver injury. It has a molecular weight of 480.38 and a chemical formula of C21H20O13.

  • Purity: 99.98%
  • Appearance: solid, off-white to light yellow
  • Molecular weight: 480.38
  • Formula: C21H20O13
  • Smiles: O=C1C(C(OC(C(C=C2O)=CC(O[C@@H]([C@@H]([C@H]3O)O)O[C@@H]([C@H]3O)CO)=C2O)=C1O)=CC(O)=C4)=C4O
  • Structure classification: flavonoids, flavonols
  • Initial source: plants, Myrtaceae, Syzygium malaccense (L.) Merr. et Perry
  • Solubility (in vitro): DMSO: 50 mg/mL (104.08 mM; needs ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Catalog No. 50-004-18743 Supplier Medchemexpress LLC Supplier No. HYN79055MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricetin 3'-glucoside is a glycoside derivative of quercetin. It significantly prevents ethanol-induced hepatotoxicity by reducing hepatic transaminase activity and inflammatory response in HepG2 cells, indicating a protective effect on alcohol-induced liver injury. It has a molecular weight of 480.38 and a chemical formula of C21H20O13.

  • Purity: 99.98%
  • Appearance: solid, off-white to light yellow
  • Molecular weight: 480.38
  • Formula: C21H20O13
  • Smiles: O=C1C(C(OC(C(C=C2O)=CC(O[C@@H]([C@@H]([C@H]3O)O)O[C@@H]([C@H]3O)CO)=C2O)=C1O)=CC(O)=C4)=C4O
  • Structure classification: flavonoids, flavonols
  • Initial source: plants, Myrtaceae, Syzygium malaccense (L.) Merr. et Perry
  • Solubility (in vitro): DMSO: 50 mg/mL (104.08 mM; needs ultrasonic; hygroscopic DMSO has a significant impact on solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.