Learn More
Medchemexpress LLC Myricetin 3-O-glucoside | 19833-12-6 | 5 MG
SDP
Catalog No. 5000418719
Encompass_Preferred
Myricetin 3-O-glucoside is a flavonol found in *Tibouchina paratropica* and Hakmeitau beans. This compound demonstrates a range of biological activities, including anti-Leishmanial, anti-inflammatory, and antimicrobial properties.
  • Demonstrates anti-Leishmanial activity
  • Demonstrates anti-inflammatory activity
  • Demonstrates antimicrobial activity
  • Exhibits anti-inflammatory effects in mouse RAW264.7 cells
Catalog No. 50-004-18719 Supplier Medchemexpress LLC Supplier No. HYN79065MG
Myricetin 3-O-glucoside is a flavonol found in *Tibouchina paratropica* and Hakmeitau beans. This compound demonstrates a range of biological activities, including anti-Leishmanial, anti-inflammatory, and antimicrobial properties.
  • Demonstrates anti-Leishmanial activity
  • Demonstrates anti-inflammatory activity
  • Demonstrates antimicrobial activity
  • Exhibits anti-inflammatory effects in mouse RAW264.7 cells