Learn More
TARGETMOL CHEMICALS INC Myricetin 3-O-glucoside 1MG
SDP
Catalog No. 502879709
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Myricetin 3-O-glucoside (Myricetin 3-O-beta-D-glucopyranoside) is a flavonol extracted from Tibouchina paratropica, exhibiting anti-leishmania, anti-inflammatory, and antibacterial activities. Purity 99.17%
Catalog No. 50-287-9709 Supplier TARGETMOL CHEMICALS INC Supplier No. TN19611MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Myricetin 3-O-glucoside (Myricetin 3-O-beta-D-glucopyranoside) is a flavonol extracted from Tibouchina paratropica, exhibiting anti-leishmania, anti-inflammatory, and antibacterial activities. Purity 99.17%