Learn More
Medchemexpress LLC Myricetin 3-O-α-L-arabinopyranoside | 132679-85-7 | 450.35 | 1 MG
SDP
Catalog No. 5000415132
Encompass_Preferred
Myricetin 3-O-α-L-arabinopyranoside is a quercetin derivative and plant flavonoid. It has antioxidant, antibacterial, and antiurease effects.
  • Inhibits A2E photooxidation-induced RPE cell death
  • Protective against retinal degeneration
  • Protects against blue light-induced damage in RPE cells and mouse models
Catalog No. 50-004-15132 Supplier Medchemexpress LLC Supplier No. HYN114991MG
Myricetin 3-O-α-L-arabinopyranoside is a quercetin derivative and plant flavonoid. It has antioxidant, antibacterial, and antiurease effects.
  • Inhibits A2E photooxidation-induced RPE cell death
  • Protective against retinal degeneration
  • Protects against blue light-induced damage in RPE cells and mouse models