Learn More
Medchemexpress LLC Myricetin 3-O-α-L-arabinopyranoside | 132679-85-7 | 450.35 | 5 MG
SDP
Catalog No. 5000415944
Encompass_Preferred
Myricetin 3-O-α-L-arabinopyranoside is a quercetin derivative and plant flavonoid. It exhibits antioxidant, antibacterial, and antiurease effects. This compound is known to inhibit A2E photooxidation-induced RPE cell death, offering protection against retinal degeneration and blue light (BL)-induced damage in RPE cells and mouse models.
  • Quercetin derivative and plant flavonoid
  • Exhibits antioxidant, antibacterial, and antiurease effects
  • Inhibits A2E photooxidation-induced RPE cell death
  • Protective against retinal degeneration
  • Safeguards against blue light (BL)-induced damage in RPE cells and mouse models
  • Intended for research use only
Catalog No. 50-004-15944 Supplier Medchemexpress LLC Supplier No. HYN114995MG
Myricetin 3-O-α-L-arabinopyranoside is a quercetin derivative and plant flavonoid. It exhibits antioxidant, antibacterial, and antiurease effects. This compound is known to inhibit A2E photooxidation-induced RPE cell death, offering protection against retinal degeneration and blue light (BL)-induced damage in RPE cells and mouse models.
  • Quercetin derivative and plant flavonoid
  • Exhibits antioxidant, antibacterial, and antiurease effects
  • Inhibits A2E photooxidation-induced RPE cell death
  • Protective against retinal degeneration
  • Safeguards against blue light (BL)-induced damage in RPE cells and mouse models
  • Intended for research use only