Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricetin-3-O-rutinoside | 41093-68-9 | 1 MG
SDP

Catalog No. 5000418153
Encompass_Preferred

Myricetin-3-O-rutinoside is a natural product that can be isolated from Picea abies. It exhibits antineuroinflammatory activity in human BV-2 cells by inhibiting LPS-induced NO production.

  • Purity: 96.98%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Molecular weight: 626.52
  • Formula: C27H30O17
  • Structure classification: Flavonoids, Flavonols
  • Initial source: Plants, Pinaceae, Picea abies (L.) H. Karsten
  • Solubility (in vitro): Ethanol: 25 mg/mL (39.90 mM; requires ultrasonic)

Catalog No. 50-004-18153 Supplier Medchemexpress LLC Supplier No. HYN79921MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricetin-3-O-rutinoside is a natural product that can be isolated from Picea abies. It exhibits antineuroinflammatory activity in human BV-2 cells by inhibiting LPS-induced NO production.

  • Purity: 96.98%
  • Appearance: Solid
  • Color: Light yellow to yellow
  • Molecular weight: 626.52
  • Formula: C27H30O17
  • Structure classification: Flavonoids, Flavonols
  • Initial source: Plants, Pinaceae, Picea abies (L.) H. Karsten
  • Solubility (in vitro): Ethanol: 25 mg/mL (39.90 mM; requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.