Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricetin-3-O-rutinoside | 41093-68-9 | 10 MG
SDP

Catalog No. 5000418154
Encompass_Preferred

Myricetin-3-O-rutinoside is a natural product isolated from Picea abies. It demonstrates antineuroinflammatory activity in human BV2 cells, inhibiting LPS-induced NO production. This product is for research use only.

  • Isolated from Picea abies
  • Antineuroinflammatory activity in human BV2 cells
  • Purity of 96.98%
  • Appearance as a solid, light yellow to yellow
  • Classified as a flavonoid and flavonol
  • Soluble in ethanol at 25 mg/mL (needs ultrasonic)

Catalog No. 50-004-18154 Supplier Medchemexpress LLC Supplier No. HYN799210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricetin-3-O-rutinoside is a natural product isolated from Picea abies. It demonstrates antineuroinflammatory activity in human BV2 cells, inhibiting LPS-induced NO production. This product is for research use only.

  • Isolated from Picea abies
  • Antineuroinflammatory activity in human BV2 cells
  • Purity of 96.98%
  • Appearance as a solid, light yellow to yellow
  • Classified as a flavonoid and flavonol
  • Soluble in ethanol at 25 mg/mL (needs ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.