Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricetin-3-O-rutinoside | 41093-68-9 | 25 MG
SDP

Catalog No. 5000421864
Encompass_Preferred

Myricetin-3-O-rutinoside is a natural product isolated from Picea abies. It has demonstrated antineuroinflammatory activity in human BV2 cells by inhibiting LPS-induced NO production after 24 hours, with an IC50 of 74.74 μM.

  • Natural product
  • Isolated from Picea abies
  • Light yellow to yellow solid appearance
  • Molecular weight: 626.52
  • Formula: C27H30O17
  • Purity: 96.98%
  • Soluble in ethanol (25 mg/mL)
  • For research use only
  • Exhibits antineuroinflammatory activity

Catalog No. 50-004-21864 Supplier Medchemexpress LLC Supplier No. 25MGHYN7992
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricetin-3-O-rutinoside is a natural product isolated from Picea abies. It has demonstrated antineuroinflammatory activity in human BV2 cells by inhibiting LPS-induced NO production after 24 hours, with an IC50 of 74.74 μM.

  • Natural product
  • Isolated from Picea abies
  • Light yellow to yellow solid appearance
  • Molecular weight: 626.52
  • Formula: C27H30O17
  • Purity: 96.98%
  • Soluble in ethanol (25 mg/mL)
  • For research use only
  • Exhibits antineuroinflammatory activity

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.