Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricetin-3-O-rutinoside | 41093-68-9 | 5 MG
SDP

Catalog No. 5000417315
Encompass_Preferred

Myricetin-3-O-rutinoside (Compound 3) is a natural product isolated from Picea abies. It demonstrates antineuroinflammatory activity in human BV2 cells, inhibiting LPS-induced NO production after 24 hours. This product is intended for research use only.

  • Isolated from Picea abies
  • Exhibits antineuroinflammatory activity
  • Purity of 96.98%
  • Molecular weight of 626.52
  • Chemical formula C27H30O17
  • Appears as a light yellow to yellow solid
  • Soluble in ethanol at 25 mg/mL (39.90 mM)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-17315 Supplier Medchemexpress LLC Supplier No. HYN79925MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricetin-3-O-rutinoside (Compound 3) is a natural product isolated from Picea abies. It demonstrates antineuroinflammatory activity in human BV2 cells, inhibiting LPS-induced NO production after 24 hours. This product is intended for research use only.

  • Isolated from Picea abies
  • Exhibits antineuroinflammatory activity
  • Purity of 96.98%
  • Molecular weight of 626.52
  • Chemical formula C27H30O17
  • Appears as a light yellow to yellow solid
  • Soluble in ethanol at 25 mg/mL (39.90 mM)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.