Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricetin-3-O-rutinoside | 41093-68-9 | 50 MG
SDP

Catalog No. 5000417629
Encompass_Preferred

Myricetin-3-O-rutinoside is a natural product isolated from Picea abies, known for its antineuroinflammatory activity. It inhibits LPS-induced NO production in human BV2 cells and is intended for research use only.

  • Isolated from Picea abies
  • Exhibits antineuroinflammatory activity in human BV2 cells
  • Inhibits LPS-induced NO production
  • Purity of 96.98%
  • Molecular weight: 626.52
  • Formula: C27H30O17
  • Appears as a light yellow to yellow solid
  • Classified as a flavonoid and flavonol
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in ethanol at 25 mg/mL (requires ultrasonic)

Catalog No. 50-004-17629 Supplier Medchemexpress LLC Supplier No. HYN799250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricetin-3-O-rutinoside is a natural product isolated from Picea abies, known for its antineuroinflammatory activity. It inhibits LPS-induced NO production in human BV2 cells and is intended for research use only.

  • Isolated from Picea abies
  • Exhibits antineuroinflammatory activity in human BV2 cells
  • Inhibits LPS-induced NO production
  • Purity of 96.98%
  • Molecular weight: 626.52
  • Formula: C27H30O17
  • Appears as a light yellow to yellow solid
  • Classified as a flavonoid and flavonol
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Soluble in ethanol at 25 mg/mL (requires ultrasonic)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.