Learn More
TARGETMOL CHEMICALS INC Myricetin 500MG
SDP
Catalog No. 502880756
Encompass_Preferred
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Myricetin (Cannabiscetin) is a natural flavonoid and MEK1 inhibitor. Myricetin has hypoglycemic, antioxidant, hepatoprotective, anti-tumor and anti-inflammatory activities. Purity 99.45%
Catalog No. 50-288-0756 Supplier TARGETMOL CHEMICALS INC Supplier No. T0579500MG
Also available in 1 mL, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Myricetin (Cannabiscetin) is a natural flavonoid and MEK1 inhibitor. Myricetin has hypoglycemic, antioxidant, hepatoprotective, anti-tumor and anti-inflammatory activities. Purity 99.45%