Learn More
Apexbio Technology LLC Myricetin 529-44-2 500mg
SDP
Catalog No. 5000089267
Encompass_Preferred
Myricetin (CAS 529-44-2) is a natural flavonoid compound isolated from Myrica rubra (Lour ) Sieb et Zucc It is known to modulate multiple cellular pathways including inhibition of protein kinases and key enzymes involved in oxidative stress and inflammation Myricetin exhibits antioxidant anti-inflammatory and anticancer properties in various in vitro and in vivo models making it a valuable probe for investigating redox biology and signal transduction Soluble in methanol ethanol acetone and ethyl acetate Myricetin should be stored tightly sealed in cool dry conditions It is widely used in biomedical research focused on molecular mechanisms of disease and therapeutic development
Catalog No. 50-000-89267 Supplier Apexbio Technology LLC Supplier No. N1850500
Myricetin (CAS 529-44-2) is a natural flavonoid compound isolated from Myrica rubra (Lour ) Sieb et Zucc It is known to modulate multiple cellular pathways including inhibition of protein kinases and key enzymes involved in oxidative stress and inflammation Myricetin exhibits antioxidant anti-inflammatory and anticancer properties in various in vitro and in vivo models making it a valuable probe for investigating redox biology and signal transduction Soluble in methanol ethanol acetone and ethyl acetate Myricetin should be stored tightly sealed in cool dry conditions It is widely used in biomedical research focused on molecular mechanisms of disease and therapeutic development