Learn More
Medchemexpress LLC Myricetin | 529-44-2 | C₁₅H₁₀O₈ | 100 MG
SDP
Catalog No. 5000374953
Encompass_Preferred
Myricetin is an analytical standard intended for research and analytical applications. This plant-derived flavonoid exhibits strong antioxidant, anticancer, antidiabetic, and anti-inflammatory activities. It is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Common plant-derived flavonoid
  • Strong antioxidant, anticancer, antidiabetic, and anti-inflammatory activities
  • Reference standard for research and analytical applications
  • Suitable for HPLC, GC, and MS experiments
Catalog No. 50-003-74953 Supplier Medchemexpress LLC Supplier No. HY15097R100MG
Myricetin is an analytical standard intended for research and analytical applications. This plant-derived flavonoid exhibits strong antioxidant, anticancer, antidiabetic, and anti-inflammatory activities. It is commonly used in qualitative, quantitative, and methodological research experiments such as HPLC, GC, and MS.
  • Common plant-derived flavonoid
  • Strong antioxidant, anticancer, antidiabetic, and anti-inflammatory activities
  • Reference standard for research and analytical applications
  • Suitable for HPLC, GC, and MS experiments