Learn More
Medchemexpress LLC Myricetin | 529-44-2 | C15H10O8 | 100 MG
SDP
Catalog No. 5000375543
Encompass_Preferred
Myricetin is a common plant-derived flavonoid known for a wide range of activities, including strong antioxidant, anticancer, antidiabetic, and anti-inflammatory properties. It is intended for research use only.
  • Exhibits scavenging activity towards radicals and ions
  • Prevents cancer cell death via apoptosis by regulating PI3K/Akt and MAPK signaling pathways
  • Displays antiphotoaging effects by quenching causative free radicals in the skin
  • Suppresses UVB-induced COX-2 expression
  • Inhibits viability of SKOV3 ovarian cancer cells in a dose-dependent manner
  • Induces DNA DSBs and ER stress, leading to apoptosis
  • Inhibits human Hsp70 by over 80%
Catalog No. 50-003-75543 Supplier Medchemexpress LLC Supplier No. HY15097100MG
Myricetin is a common plant-derived flavonoid known for a wide range of activities, including strong antioxidant, anticancer, antidiabetic, and anti-inflammatory properties. It is intended for research use only.
  • Exhibits scavenging activity towards radicals and ions
  • Prevents cancer cell death via apoptosis by regulating PI3K/Akt and MAPK signaling pathways
  • Displays antiphotoaging effects by quenching causative free radicals in the skin
  • Suppresses UVB-induced COX-2 expression
  • Inhibits viability of SKOV3 ovarian cancer cells in a dose-dependent manner
  • Induces DNA DSBs and ER stress, leading to apoptosis
  • Inhibits human Hsp70 by over 80%