Learn More
Medchemexpress LLC Myricetin | 529-44-2 | C15H10O8 | 200 MG
SDP
Catalog No. 5000374992
Encompass_Preferred
Myricetin is a plant-derived flavonoid known for its strong antioxidant, anticancer, antidiabetic, and anti-inflammatory properties. It exhibits diverse cellular effects, including radical scavenging activity and the ability to modulate various signaling pathways, making it suitable for a wide range of research applications.
  • Exhibits scavenging activity against various radicals and ions.
  • Prevents cancer cell death by apoptosis through regulation of PI3K/Akt and MAPK signaling pathways.
  • Displays antiphotoaging effects by quenching free radicals in the skin and suppressing UVB-induced COX-2 expression.
  • Inhibits human Hsp70 with notable IC50 values.
  • Causes inhibition of rabbit platelet aggregation induced by ADP, arachidonic acid, collagen, and PAF.
Catalog No. 50-003-74992 Supplier Medchemexpress LLC Supplier No. HY15097200MG
Myricetin is a plant-derived flavonoid known for its strong antioxidant, anticancer, antidiabetic, and anti-inflammatory properties. It exhibits diverse cellular effects, including radical scavenging activity and the ability to modulate various signaling pathways, making it suitable for a wide range of research applications.
  • Exhibits scavenging activity against various radicals and ions.
  • Prevents cancer cell death by apoptosis through regulation of PI3K/Akt and MAPK signaling pathways.
  • Displays antiphotoaging effects by quenching free radicals in the skin and suppressing UVB-induced COX-2 expression.
  • Inhibits human Hsp70 with notable IC50 values.
  • Causes inhibition of rabbit platelet aggregation induced by ADP, arachidonic acid, collagen, and PAF.