Learn More
Medchemexpress LLC Myricetin | 529-44-2 | C15H10O8 | 25 MG
SDP
Catalog No. 5000375953
Encompass_Preferred
Myricetin is a plant-derived flavonoid with strong antioxidant, anticancer, antidiabetic, and anti-inflammatory properties. It exhibits scavenging activity against radicals and ions, prevents cancer cell death via apoptosis through PI3K/Akt and MAPK signaling, and shows antiphotoaging effects by quenching free radicals in the skin. It also inhibits ovarian cancer cell viability and human Hsp70.
  • Strong antioxidant, anticancer, antidiabetic, and anti-inflammatory activities
  • Scavenges various radicals and ions
  • Prevents cancer cell death via apoptosis
  • Exhibits antiphotoaging effects in skin
  • Suppresses UVB-induced COX-2 expression
  • Inhibits UVB-induced initiation of activator protein-1 and NF-κβ
  • Inhibits Fyn kinase activity
  • Inhibits viability of SKOV3 ovarian cancer cells
  • Induces DNA DSBs and ER stress leading to apoptosis
  • Inhibits human Hsp70
Catalog No. 50-003-75953 Supplier Medchemexpress LLC Supplier No. 25MGHY15097R
Myricetin is a plant-derived flavonoid with strong antioxidant, anticancer, antidiabetic, and anti-inflammatory properties. It exhibits scavenging activity against radicals and ions, prevents cancer cell death via apoptosis through PI3K/Akt and MAPK signaling, and shows antiphotoaging effects by quenching free radicals in the skin. It also inhibits ovarian cancer cell viability and human Hsp70.
  • Strong antioxidant, anticancer, antidiabetic, and anti-inflammatory activities
  • Scavenges various radicals and ions
  • Prevents cancer cell death via apoptosis
  • Exhibits antiphotoaging effects in skin
  • Suppresses UVB-induced COX-2 expression
  • Inhibits UVB-induced initiation of activator protein-1 and NF-κβ
  • Inhibits Fyn kinase activity
  • Inhibits viability of SKOV3 ovarian cancer cells
  • Induces DNA DSBs and ER stress leading to apoptosis
  • Inhibits human Hsp70