Learn More
Medchemexpress LLC Myricetin | 529-44-2 | C15H10O8 | 500 MG
SDP
Catalog No. 5000375481
Encompass_Preferred
Myricetin is a naturally occurring plant-derived flavonoid known for its diverse biological activities, including potent anti-oxidant, anticancer, antidiabetic, and anti-inflammatory properties. It operates through various mechanisms, influencing key cellular pathways and demonstrating potential therapeutic applications.
  • Scavenges various radicals and ions
  • Prevents cancer cell death through apoptosis regulation
  • Exhibits antiphotoaging effects by neutralizing free radicals in the skin
  • Inhibits viability of ovarian cancer cells
  • Inhibits human Hsp70
  • Shows potential in reducing tumor growth and metastatic spread
  • Inhibits platelet aggregation
Catalog No. 50-003-75481 Supplier Medchemexpress LLC Supplier No. HY15097500MG
Myricetin is a naturally occurring plant-derived flavonoid known for its diverse biological activities, including potent anti-oxidant, anticancer, antidiabetic, and anti-inflammatory properties. It operates through various mechanisms, influencing key cellular pathways and demonstrating potential therapeutic applications.
  • Scavenges various radicals and ions
  • Prevents cancer cell death through apoptosis regulation
  • Exhibits antiphotoaging effects by neutralizing free radicals in the skin
  • Inhibits viability of ovarian cancer cells
  • Inhibits human Hsp70
  • Shows potential in reducing tumor growth and metastatic spread
  • Inhibits platelet aggregation