Learn More
Medchemexpress LLC Myricetin | 529-44-2 | MFCD00006827 | 98.5% | 318.24 g/mol | C15H10O8 | 10 MG
SDP
Catalog No. 5000287149
Encompass_Preferred
Myricetin is an analytical reference standard of myricetin (CAS 529-44-2), a plant-derived flavonoid used as a reference material in analytical and research applications. Supplied as a high-purity solid suitable for HPLC, method validation, and quantitative analysis.
  • Analytical reference standard for HPLC and method validation.
  • High purity: 98.5% assay.
  • Provided in small, controlled pack sizes for accurate weighing.
  • Known molecular weight and formula for identification and calibration.
  • Intended for research and analytical applications.
Catalog No. 50-002-87149 Supplier Medchemexpress LLC Supplier No. HY15097R10MG
Myricetin is an analytical reference standard of myricetin (CAS 529-44-2), a plant-derived flavonoid used as a reference material in analytical and research applications. Supplied as a high-purity solid suitable for HPLC, method validation, and quantitative analysis.
  • Analytical reference standard for HPLC and method validation.
  • High purity: 98.5% assay.
  • Provided in small, controlled pack sizes for accurate weighing.
  • Known molecular weight and formula for identification and calibration.
  • Intended for research and analytical applications.