Learn More
Selleck Chemical LLC Myricetin 100mg 529-44-2 Cannabiscetin
SDP
Catalog No. 505750
Encompass_Preferred
Myricetin, a natural flavonoid with antioxidant and anti tumor properties, is a novel inhibitor of MEK1 activity and transformation of JB6 P+ mouse epidermal cells. It also inhibits PI3K? with Kd of 0.17 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-575-0 Supplier Selleck Chemical LLC Supplier No. S2326100MG
Myricetin, a natural flavonoid with antioxidant and anti tumor properties, is a novel inhibitor of MEK1 activity and transformation of JB6 P+ mouse epidermal cells. It also inhibits PI3K? with Kd of 0.17 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.