Learn More
Abcam Myricetin, irreversible Tr x R inhibitor, 5MG
Catalog No. 502707338
Encompass_Preferred
MW 318.24 g/mol, Purity >97%. Directly inhibits MEK, JAK1, Akt and MKK4 kinase activity. Able to inhibit cytochrome P450 3A4 and 2C9 (IC₅₀ = 7.81 and 13.5 μM, respectively). .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7338 Supplier Abcam Supplier No. AB1207215MG
MW 318.24 g/mol, Purity >97%. Directly inhibits MEK, JAK1, Akt and MKK4 kinase activity. Able to inhibit cytochrome P450 3A4 and 2C9 (IC₅₀ = 7.81 and 13.5 μM, respectively). .
The product is subject to the following: Abcam Restricted Use Statement