Learn More
Cayman Chemical Myricetin Natural Products Ext
SDP
Supplier:  Cayman Chemical 10012600100 MG
Encompass_Preferred
A flavonoid compound that acts as a powerful antioxidant; inhibits TBARS formation with an IC50 value of 6.34 µM; blocks oxLDL uptake by U937-derived macrophages at 20 µM; demonstrates potent chemopreventative potential by binding JAK1/STAT3 to inhibit neoplastic transformation of murine JB6 Pt cells
Catalog No. 50-288-8922