Learn More
Cayman Chemical Myricetin, Purity: ≥98%, CAS: 529-44-2, 25 mg
SDP
Catalog No. 502890224
Encompass_Preferred
A flavonoid compound that acts as a powerful antioxidant; inhibits TBARS formation with an IC50 value of 6.34 µM; blocks oxLDL uptake by U937-derived macrophages at 20 µM; demonstrates potent chemopreventative potential by binding JAK1/STAT3 to inhibit neoplastic transformation of murine JB6 Pt cells
Catalog No. 50-289-0224 Supplier Cayman Chemical Supplier No. 1001260025 MG
A flavonoid compound that acts as a powerful antioxidant; inhibits TBARS formation with an IC50 value of 6.34 µM; blocks oxLDL uptake by U937-derived macrophages at 20 µM; demonstrates potent chemopreventative potential by binding JAK1/STAT3 to inhibit neoplastic transformation of murine JB6 Pt cells