Learn More
Medchemexpress LLC Myricetin (Standard) | 529-44-2 | MFCD00006827 | C₁₅H₁₀O₈ | 50 MG
SDP
Catalog No. 5000375483
Encompass_Preferred
Myricetin (Standard) is an analytical standard and a common plant-derived flavonoid, also identified as an endogenous human gut microbiota metabolite. It is extensively researched for its strong anti-oxidant, anticancer, antidiabetic, and anti-inflammatory properties, making it ideal for various research and analytical applications.
  • For research and analytical applications
  • Plant-derived flavonoid and gut microbiota metabolite
  • Strong anti-oxidant, anticancer, antidiabetic, anti-inflammatory activities
  • Analytical standard grade for reference assays
  • Supports qualitative, quantitative, and methodological research
  • Compatible with HPLC, GC, and MS techniques
  • Solid, light yellow to yellow appearance
Catalog No. 50-003-75483 Supplier Medchemexpress LLC Supplier No. HY15097R50MG
Myricetin (Standard) is an analytical standard and a common plant-derived flavonoid, also identified as an endogenous human gut microbiota metabolite. It is extensively researched for its strong anti-oxidant, anticancer, antidiabetic, and anti-inflammatory properties, making it ideal for various research and analytical applications.
  • For research and analytical applications
  • Plant-derived flavonoid and gut microbiota metabolite
  • Strong anti-oxidant, anticancer, antidiabetic, anti-inflammatory activities
  • Analytical standard grade for reference assays
  • Supports qualitative, quantitative, and methodological research
  • Compatible with HPLC, GC, and MS techniques
  • Solid, light yellow to yellow appearance