Learn More
Medchemexpress LLC Myricitrin | 17912-87-7 | 99.8% | 464.38 | 25 MG
SDP
Catalog No. 5000430089
Encompass_Preferred
Myricitrin, a naturally occurring flavonoid, acts as an orally active nitric oxide (NO) and PKC inhibitor. It exhibits central nervous system activity, including anxiolytic-like effects. Myricitrin also possesses antioxidant, anti-inflammatory, antifibrotic, and anti-malarial properties.
  • Orally active NO and PKC inhibitor.
  • Central nervous system activity (anxiolytic-like action).
  • Antioxidant effects (radical scavenger, metal-ion chelator).
  • Anti-inflammatory effects.
  • Antifibrotic effects.
  • Anti-malarial effects (inhibits *Plasmodium falciparum* proliferation).
  • Inhibits aldose reductase activity and low-density lipoprotein oxidation.
  • Exhibits scavenging activity against DPPH, NO radicals, and H2O2.
  • Inhibits TNFα production and enhances RANTES production in LPS-stimulated Raw264.7 cells.
Catalog No. 50-004-30089 Supplier Medchemexpress LLC Supplier No. 25MGHYN0152
Myricitrin, a naturally occurring flavonoid, acts as an orally active nitric oxide (NO) and PKC inhibitor. It exhibits central nervous system activity, including anxiolytic-like effects. Myricitrin also possesses antioxidant, anti-inflammatory, antifibrotic, and anti-malarial properties.
  • Orally active NO and PKC inhibitor.
  • Central nervous system activity (anxiolytic-like action).
  • Antioxidant effects (radical scavenger, metal-ion chelator).
  • Anti-inflammatory effects.
  • Antifibrotic effects.
  • Anti-malarial effects (inhibits *Plasmodium falciparum* proliferation).
  • Inhibits aldose reductase activity and low-density lipoprotein oxidation.
  • Exhibits scavenging activity against DPPH, NO radicals, and H2O2.
  • Inhibits TNFα production and enhances RANTES production in LPS-stimulated Raw264.7 cells.