Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myricitrin | 17912-87-7 | MFCD00016930 | 99.8% | 464.38 | C21H20O12 | 10 MG
SDP

Catalog No. 5000295824
Encompass_Preferred

Myricitrin is a naturally occurring flavonoid used in research as a nitric oxide (NO) and protein kinase C (PKC) inhibitor. It is supplied in small-mass vials for laboratory use and includes solubility and storage guidance for in vitro and in vivo formulation.

  • High purity (99.76%) suitable for analytical and in vitro studies.
  • Molecular weight 464.38 and formula C21H20O12.
  • Available as 10 mg vials.
  • Soluble in DMSO (100 mg/mL; may require ultrasonic) and formulation solubility ≥ 2.5 mg/mL.
  • Store at 4°C, protect from light; in solvent store at -80°C up to 6 months.

Catalog No. 50-002-95824 Supplier Medchemexpress LLC Supplier No. HYN015210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myricitrin is a naturally occurring flavonoid used in research as a nitric oxide (NO) and protein kinase C (PKC) inhibitor. It is supplied in small-mass vials for laboratory use and includes solubility and storage guidance for in vitro and in vivo formulation.

  • High purity (99.76%) suitable for analytical and in vitro studies.
  • Molecular weight 464.38 and formula C21H20O12.
  • Available as 10 mg vials.
  • Soluble in DMSO (100 mg/mL; may require ultrasonic) and formulation solubility ≥ 2.5 mg/mL.
  • Store at 4°C, protect from light; in solvent store at -80°C up to 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.