Learn More
Cayman Chemical Myrislignan, Purity: ≥98%, CAS: 171485-39-5, 50 mg
SDP
Catalog No. 502947619
Encompass_Preferred
A lignan with diverse biological activities; inhibits hypocotyl or root growth of cress (IC50s = 429 and 517 µM, respectively); inhibits production of NO, increases in iNOS and COX-2 levels, and release of TNF-α and IL-6 in LPS-stimulated RAW 264.7 cells at 50 µg/ml; inhibits proliferation of, and induces apoptosis in, A549 lung cancer cells; reduces tumor growth in an A549 lung cancer mouse xenograft model at 3, 5, and 8 mg/kg; decreases parasite burden in the liver, brain, and spleen in a mouse model of T. gondii infection at 100 mg/kg
Catalog No. 50-294-7619 Supplier Cayman Chemical Supplier No. 3375950 MG
A lignan with diverse biological activities; inhibits hypocotyl or root growth of cress (IC50s = 429 and 517 µM, respectively); inhibits production of NO, increases in iNOS and COX-2 levels, and release of TNF-α and IL-6 in LPS-stimulated RAW 264.7 cells at 50 µg/ml; inhibits proliferation of, and induces apoptosis in, A549 lung cancer cells; reduces tumor growth in an A549 lung cancer mouse xenograft model at 3, 5, and 8 mg/kg; decreases parasite burden in the liver, brain, and spleen in a mouse model of T. gondii infection at 100 mg/kg