Learn More
Medchemexpress LLC Myristamidopropyl di 250mg
SDP
Catalog No. 5000204518
Encompass_Preferred
Myristamidopropyl dimethylamine (MAPD) is an antimicrobial agent (including against bacteria and fungi) and an insecticide exhibiting inhibitory activity against Pseudomonas aeruginosa Staphylococcus aureus Candida albicans Fusarium solani and Acanthamoeba polyphaga Myristamidopropyl dimethylamine can be used in research on microbial-induced keratitis [1]
Catalog No. 50-002-04518 Supplier Medchemexpress LLC Supplier No. 250MGHYW099582
Myristamidopropyl dimethylamine (MAPD) is an antimicrobial agent (including against bacteria and fungi) and an insecticide exhibiting inhibitory activity against Pseudomonas aeruginosa Staphylococcus aureus Candida albicans Fusarium solani and Acanthamoeba polyphaga Myristamidopropyl dimethylamine can be used in research on microbial-induced keratitis [1]