Learn More
Cayman Chemical MyrIstelaIdIc AcId 10mg
SDP
Catalog No. 502987659
Encompass_Preferred
A monounsaturated ω-5 fatty acid; the trans isomer of myristoleic acid; converted to myristelaidoyl CoA at a faster rate than myristoleic acid to myristoleoyl CoA
Catalog No. 50-298-7659 Supplier Cayman Chemical Supplier No. 2236110 MG
A monounsaturated ω-5 fatty acid; the trans isomer of myristoleic acid; converted to myristelaidoyl CoA at a faster rate than myristoleic acid to myristoleoyl CoA