Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Myristelaidic acid | 50286-30-1 | 99.8% | 226.36 | C14H26O2 | 1MG
SDP

Catalog No. 5000238429
Encompass_Preferred

Myristelaidic acid is a 14-carbon trans monounsaturated fatty acid (trans-9-tetradecenoic acid) supplied as a high-purity research reagent for use as a reference standard and in lipid, metabolic, and analytical studies.

  • Cas number 50286-30-1
  • Molecular formula C14H26O2
  • Molecular weight 226.36 g/mol
  • Purity 99.8%
  • Physical form liquid
  • Soluble in DMSO (≥ 100 mg/mL) and ethanol (≥ 10 mg/mL)
  • Available in 1 mg, 5 mg, and 10 mg quantities

Catalog No. 50-002-38429 Supplier Medchemexpress LLC Supplier No. HY1241411MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Myristelaidic acid is a 14-carbon trans monounsaturated fatty acid (trans-9-tetradecenoic acid) supplied as a high-purity research reagent for use as a reference standard and in lipid, metabolic, and analytical studies.

  • Cas number 50286-30-1
  • Molecular formula C14H26O2
  • Molecular weight 226.36 g/mol
  • Purity 99.8%
  • Physical form liquid
  • Soluble in DMSO (≥ 100 mg/mL) and ethanol (≥ 10 mg/mL)
  • Available in 1 mg, 5 mg, and 10 mg quantities

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.