Learn More
Medchemexpress LLC Myristelaidic acid | 50286-30-1 | 99.8% | 226.36 g/mol | C14H26O2 | 5MG
SDP
Catalog No. 5000236970
Encompass_Preferred
Myristelaidic acid (trans-9-tetradecenoic acid) is the trans isomer of myristoleic acid - a 14-carbon trans fatty acid found in many animal fats. It is supplied as a research-grade reagent for biochemical and analytical studies.
  • 14-carbon trans fatty acid; molecular formula C14H26O2, MW 226.36 g/mol.
  • High purity (99.8%).
  • Available in small analytical pack sizes: 1 mg, 5 mg, 10 mg.
  • Provided with product data sheet and safety data sheets.
  • Intended for research and analytical applications.
Catalog No. 50-002-36970 Supplier Medchemexpress LLC Supplier No. HY1241415MG
Add to Cart
Add to Cart
Myristelaidic acid (trans-9-tetradecenoic acid) is the trans isomer of myristoleic acid - a 14-carbon trans fatty acid found in many animal fats. It is supplied as a research-grade reagent for biochemical and analytical studies.
  • 14-carbon trans fatty acid; molecular formula C14H26O2, MW 226.36 g/mol.
  • High purity (99.8%).
  • Available in small analytical pack sizes: 1 mg, 5 mg, 10 mg.
  • Provided with product data sheet and safety data sheets.
  • Intended for research and analytical applications.