Learn More
Medchemexpress LLC Myristelaidic acid | 50286-30-1 | MFCD00133547 | 99.8% | 226.36 | C14H26O2 | 10MG
SDP
Catalog No. 5000238422
Encompass_Preferred
Myristelaidic acid (trans-9-tetradecenoic acid) is a 14-carbon trans monounsaturated fatty acid supplied for research use. It is the trans isomer of myristoleic acid and occurs in many animal fats. The product is provided as a purified analytical reagent; consult the certificate of analysis and safety data sheet for purity, handling, and storage recommendations.
Catalog No. 50-002-38422 Supplier Medchemexpress LLC Supplier No. HY12414110MG
Add to Cart
Add to Cart
Myristelaidic acid (trans-9-tetradecenoic acid) is a 14-carbon trans monounsaturated fatty acid supplied for research use. It is the trans isomer of myristoleic acid and occurs in many animal fats. The product is provided as a purified analytical reagent; consult the certificate of analysis and safety data sheet for purity, handling, and storage recommendations.