Learn More
Cayman Chemical Myristelaidic Acid, Purity: ≥98%, CAS: 50286-30-1, 50 mg
SDP
Catalog No. 502987942
Encompass_Preferred
A monounsaturated ω-5 fatty acid; the trans isomer of myristoleic acid; converted to myristelaidoyl CoA at a faster rate than myristoleic acid to myristoleoyl CoA
Catalog No. 50-298-7942 Supplier Cayman Chemical Supplier No. 2236150 MG
A monounsaturated ω-5 fatty acid; the trans isomer of myristoleic acid; converted to myristelaidoyl CoA at a faster rate than myristoleic acid to myristoleoyl CoA