Learn More
Cayman Chemical Myristic Acid-d27 methyl ester, Purity: ≥99% deuterated forms (d1-d27), CAS: 434349-93-6, 25 mg
SDP
Catalog No. 502938597
Encompass_Preferred
An internal standard for the quantification of myristic acid methyl ester by GC- or LC-MS
Catalog No. 50-293-8597 Supplier Cayman Chemical Supplier No. 2859425 MG
An internal standard for the quantification of myristic acid methyl ester by GC- or LC-MS