Learn More
Cayman Chemical MyrIstIc AcId 10g
SDP
Catalog No. 502953758
Encompass_Preferred
A saturated fatty acid; incorporated into myristoyl-CoA and transferred by N-myristoyltransferase to the N-terminal glycine of certain proteins
Catalog No. 50-295-3758 Supplier Cayman Chemical Supplier No. 1335110 G
A saturated fatty acid; incorporated into myristoyl-CoA and transferred by N-myristoyltransferase to the N-terminal glycine of certain proteins