Learn More
Cayman Chemical Myristic Acid-13C, Purity: ≥95%, CAS: 57677-52-8, 1 g
SDP
Catalog No. 502989100
Encompass_Preferred
An internal standard for the quantification of myristic acid by GC- or LC-MS
Catalog No. 50-298-9100 Supplier Cayman Chemical Supplier No. 294631 G
An internal standard for the quantification of myristic acid by GC- or LC-MS