Learn More
Cayman Chemical Myristic Acid-13C, Purity: ≥95%, CAS: 57677-52-8, 500 mg
SDP
Catalog No. 502992540
Encompass_Preferred
An internal standard for the quantification of myristic acid by GC- or LC-MS
Catalog No. 50-299-2540 Supplier Cayman Chemical Supplier No. 29463500 MG
An internal standard for the quantification of myristic acid by GC- or LC-MS