Learn More
Medchemexpress LLC HY-N2041 1g Medchemexpress, Myristic acid CAS:544-63-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN20411G
Encompass_Preferred
Medchemexpress, HY-N2041 1g Myristic acid CAS:544-63-8 Myristic acid is a saturated 14-carbon fatty acid occurring in most animal and vegetable fats, particularly butterfat and coconut, palm, and nutmeg oils. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3877