Learn More
Cayman Chemical Myristic Acid, Purity: ≥98%, CAS: 544-63-8, 5 g
SDP
Catalog No. 502954828
Encompass_Preferred
A saturated fatty acid; incorporated into myristoyl-CoA and transferred by N-myristoyltransferase to the N-terminal glycine of certain proteins
Catalog No. 50-295-4828 Supplier Cayman Chemical Supplier No. 133515 G
A saturated fatty acid; incorporated into myristoyl-CoA and transferred by N-myristoyltransferase to the N-terminal glycine of certain proteins