Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Myristic acid, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014556 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4556 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4556 Supplier MilliporeSigma Supelco Supplier No. 700795G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Myristic acid is an important bioactive component of Suanzaoren seed, which is one of the most commonly used Chinese medicine with therapeutic properties such as hypotensive, antihypoxia, hypnotic-sedative, antihyperlipidemia, and hypothermic effects.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 544-63-8
Flash Point >113°C
Physical Form Neat
Linear Formula CH3(CH2)12 COOH
MDL Number MFCD00002744
Melting Point 52°C to 54°C; 53°C to 56°C
Boiling Point 250°C (100 mmHg)
Synonym 1-Tridecanecarboxylic acid; C14:0; NSC 5028; Tetradecanoic acid
Type Analytical Standard
Quantity 5 g
Formula Weight 228.37
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.