Learn More
Cayman Chemical MyrIstIc AcId Alkyn 100ug
SDP
Catalog No. 502952900
Encompass_Preferred
A form of the 14-carbon myristic acid with an ω-terminal alkyne group; can be used in linking reactions utilizing click chemistry
Catalog No. 50-295-2900 Supplier Cayman Chemical Supplier No. 13267100 UG
A form of the 14-carbon myristic acid with an ω-terminal alkyne group; can be used in linking reactions utilizing click chemistry