Learn More
Cayman Chemical Myristic Acid Alkyne, Purity: ≥98%, CAS: 82909-47-5, 1 mg
SDP
Catalog No. 502952788
Encompass_Preferred
A form of the 14-carbon myristic acid with an ω-terminal alkyne group; can be used in linking reactions utilizing click chemistry
Catalog No. 50-295-2788 Supplier Cayman Chemical Supplier No. 132671 MG
A form of the 14-carbon myristic acid with an ω-terminal alkyne group; can be used in linking reactions utilizing click chemistry