Learn More
Cayman Chemical MyrIstIc AcId Alkyn 500ug
SDP
Catalog No. 502952859
Encompass_Preferred
A form of the 14-carbon myristic acid with an ω-terminal alkyne group; can be used in linking reactions utilizing click chemistry
Catalog No. 50-295-2859 Supplier Cayman Chemical Supplier No. 13267500 UG
A form of the 14-carbon myristic acid with an ω-terminal alkyne group; can be used in linking reactions utilizing click chemistry