Learn More
Cayman Chemical Myristic Acid-d2, Purity: ≥99% deuterated forms (d1-d2), CAS: 30719-21-2, 250 mg
SDP
Catalog No. 502990612
Encompass_Preferred
An internal standard for the quantification of myristic acid by GC- or LC-MS
Catalog No. 50-299-0612 Supplier Cayman Chemical Supplier No. 29411250 MG
An internal standard for the quantification of myristic acid by GC- or LC-MS