Learn More
Cayman Chemical Myristic Acid-d27, Purity: ≥99% deuterated forms (d1-d27), CAS: 60658-41-5, 100 mg
SDP
Catalog No. 502896089
Encompass_Preferred
An internal standard for the quantification of myristic acid by GC- or LC-MS
Catalog No. 50-289-6089 Supplier Cayman Chemical Supplier No. 9003317100 MG
An internal standard for the quantification of myristic acid by GC- or LC-MS