Learn More
Medchemexpress LLC Myristic acid-d27 | 60658-41-5 | 255.54 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYN2041S100MG
Encompass_Preferred
Myristic acid-d27 is the deuterium labeled Myristic acid. Myristic acid is a saturated 14-carbon fatty acid occurring in most animal and vegetable fats, particularly butterfat and coconut, palm, and nutmeg oils. It is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-15223