Learn More
Medchemexpress LLC Myristic acid-d27 | 60658-41-5 | 98.8% | 255.54 | C14HD27O2 | 10 MG
SDP
Catalog No. 5000296621
Encompass_Preferred
Myristic acid-d27 is the deuterium-labeled isotopologue of myristic (tetradecanoic) acid supplied for analytical and tracer applications. It is provided as a solid and is suitable as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled (d27) for use as a tracer or internal standard.
  • Saturated 14-carbon fatty acid structure compatible with lipid analyses.
  • High reported purity suitable for quantitative assays.
  • Provided as a 10 mg solid pack for small-scale analytical use.
  • Stable under suggested storage conditions to maintain isotopic integrity.
Catalog No. 50-002-96621 Supplier Medchemexpress LLC Supplier No. HYN2041S10MG
Myristic acid-d27 is the deuterium-labeled isotopologue of myristic (tetradecanoic) acid supplied for analytical and tracer applications. It is provided as a solid and is suitable as an internal standard for quantitative analysis by NMR, GC-MS, and LC-MS.
  • Deuterium-labeled (d27) for use as a tracer or internal standard.
  • Saturated 14-carbon fatty acid structure compatible with lipid analyses.
  • High reported purity suitable for quantitative assays.
  • Provided as a 10 mg solid pack for small-scale analytical use.
  • Stable under suggested storage conditions to maintain isotopic integrity.