Learn More
Medchemexpress LLC Myristic acid-d5 | 327077-03-2 | MFCD03428167 | 98.0% | 233.40 | C14H23D5O2 | 10 MG
SDP
Catalog No. 5000296390
Encompass_Preferred
Myristic acid-d5 is a deuterium-labeled derivative of myristic acid used as a stable isotope tracer and internal standard for analytical chemistry. Supplied as a white to off-white solid, it is suitable for quantitative analysis by NMR, GC-MS, and LC-MS and intended for research use only.
  • Stable isotope labeled (d5) tracer for analytical methods.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity: 98.0%.
  • Supplied as a solid in a 10 MG package.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).
Catalog No. 50-002-96390 Supplier Medchemexpress LLC Supplier No. HYN2041S610MG
Myristic acid-d5 is a deuterium-labeled derivative of myristic acid used as a stable isotope tracer and internal standard for analytical chemistry. Supplied as a white to off-white solid, it is suitable for quantitative analysis by NMR, GC-MS, and LC-MS and intended for research use only.
  • Stable isotope labeled (d5) tracer for analytical methods.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity: 98.0%.
  • Supplied as a solid in a 10 MG package.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months).