Learn More
Cayman Chemical Myristic Acid-d7, Purity: ≥99% deuterated forms (d1-d7), 10 mg
SDP
Catalog No. 502942577
Encompass_Preferred
An internal standard for the quantification of myristic acid by GC- or LC-MS
Catalog No. 50-294-2577 Supplier Cayman Chemical Supplier No. 3985910 MG
An internal standard for the quantification of myristic acid by GC- or LC-MS